missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-propylimidazolium Iodide 97.0+%, TCI America™
Supplier: TCI America M14405G
Specifications
| 1-Methyl-3-propylimidazolium Iodide | |
| C7H13IN2 | |
| 5 g | |
| IVCMUVGRRDWTDK-UHFFFAOYSA-M | |
| 3-methyl-1-propyl-1H-imidazol-3-ium iodide | |
| 12971008 | |
| ≥97.0% (HPLC,T) |
| 119171-18-5 | |
| MFCD08276385 | |
| 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide | |
| [I-].CCCN1C=C[N+](C)=C1 | |
| 252.10 | |
| 252.10 | |
| Liquid |
Chemical Identifiers
| 119171-18-5 | |
| 252.10 | |
| IVCMUVGRRDWTDK-UHFFFAOYSA-M | |
| 12971008 | |
| [I-].CCCN1C=C[N+](C)=C1 |
| C7H13IN2 | |
| MFCD08276385 | |
| 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide | |
| 3-methyl-1-propyl-1H-imidazol-3-ium iodide |
Safety and Handling
TSCA : No