missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-Toluenesulfonate 95.0+%, TCI America™

Catalog No. p-7133161
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23211G 1 g
M23215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23211G Supplier TCI America Supplier No. M23211G

Chemical Identifiers

CAS 1352947-63-7
Molecular Formula C18H28N2O3S2
Molecular Weight (g/mol) 384.553
InChI Key OVUBPNDERNKKQU-UHFFFAOYSA-M
PubChem CID 66552686
IUPAC Name 4-methylbenzenesulfonate;1-methyl-3-(6-methylsulfanylhexyl)imidazol-1-ium
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CN(C=C1)CCCCCCSC

Specifications

Color White
Quantity 1 g
Formula Weight 384.55
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-Toluenesulfonate