accessibility menu, dialog, popup

UserName

CH3SOOOH3CNNSCH3

CAS RN 1352947-63-7

IUPAC Name: 3-methyl-1-[6-(methylsulfanyl)hexyl]-1H-imidazol-3-ium 4-methylbenzene-1-sulfonate
Synonyms: 1-methyl-3-[6-(methylsulfanyl)hexyl]imidazol-1-ium tosylate
Molecular Weight (g/mol): 384.55
Molecular Formula: C18H28N2O3S2
InChi Key: OVUBPNDERNKKQU-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.CSCCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results