Learn More
1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%
CAS Number-872-50-4, 4 x 1L, Colorless liquid, 106420, 204 deg.C, 1.033, 212-828-1, 86 deg.C (187 deg.F), 99.13, HPLC, H227-H315-H319-H335-H360D-H373, P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c, 270 deg.C (518 deg.F) | CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Supplier: Thermo Scientific Chemicals 038986K7
Description
1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Methyl-2-pyrrolidinone | |
| 872-50-4 | |
| Colorless liquid | |
| 204°C | |
| 86°C (187°F) | |
| Amine-like | |
| 1.4705 | |
| MFCD00003193 | |
| 106420 | |
| SECXISVLQFMRJM-UHFFFAOYSA-N | |
| 1-methylpyrrolidin-2-one | |
| 99.13 |
| °C to 24°C | |
| Colorless | |
| 1.033 | |
| 4 L | |
| 270°C (518°F) | |
| Liquid | |
| C5H9NO | |
| 0.29 hPa at 20°C | |
| Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons. | |
| CN1CCCC1=O | |
| 99.13 | |
| HPLC |
Chemical Identifiers
| 872-50-4 | |
| 99.13 | |
| SECXISVLQFMRJM-UHFFFAOYSA-N | |
| CN1CCCC1=O |
| C5H9NO | |
| MFCD00003193 | |
| 1-methylpyrrolidin-2-one |
Safety and Handling
P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335-H360D-H373
EINECSNumber : 212-828-1
RTECSNumber : UY5790000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only