missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%

Catalog No. p-3256802
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38986K2 1 L
AA38986M1 4 x 1 L
AA38986K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA38986K2 Supplier Thermo Scientific Chemicals Supplier No. 038986K2
Only null left

CAS Number-872-50-4, 4 x 1L, Colorless liquid, 106420, 204 deg.C, 1.033, 212-828-1, 86 deg.C (187 deg.F), 99.13, HPLC, H227-H315-H319-H335-H360D-H373, P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c, 270 deg.C (518 deg.F) | CAS: 872-50-4 | C5H9NO | 99.13 g/mol

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point °C to 24°C
Color Colorless
Appearance Colorless liquid
Density 1.033 g/cm3 at 20°C
Boiling Point 204°C
Quantity 1 L
Flash Point 86°C (187°F)
Ignition Point 270°C (518°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.4705
Vapor Pressure 0.29 hPa at 20°C
Beilstein 106420
Solubility Information Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Formula Weight 99.13
Grade HPLC
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
Hazard Category H227-H315-H319-H335-H360D-H373
Precautionary Statement P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-828-1
RTECSNumber UY5790000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only