missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1-phenylhydrazine 95.0+%, TCI America™

$115.22 - $337.41

Chemical Identifiers

CAS 618-40-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00007627
InChI Key MWOODERJGVWYJE-UHFFFAOYSA-N
Synonym 1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
PubChem CID 36299
IUPAC Name 1-methyl-1-phenylhydrazine
SMILES CN(C1=CC=CC=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M03995G View Documents Tci America
M03995G
5g Each for $115.22
 
M039925G View Documents Tci America
M039925G
25g Each for $337.41
 
Specifications

Chemical Identifiers

618-40-6
122.171
MWOODERJGVWYJE-UHFFFAOYSA-N
36299
CN(C1=CC=CC=C1)N
C7H10N2
MFCD00007627
1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
1-methyl-1-phenylhydrazine

Specifications

109°C
5g
C7H10N2
1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
CN(C1=CC=CC=C1)N
122.171
122.17
≥95.0% (GC)
Undesignated
618-40-6
MFCD00007627
MWOODERJGVWYJE-UHFFFAOYSA-N
1-methyl-1-phenylhydrazine
36299
Clear Liquid at 20°C
1-Methyl-1-phenylhydrazine
Safety and Handling

Safety and Handling

RTECSNumber : MV7712000

TSCA : Yes