missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-phenylhydrazine, 97%

Catalog No. AAB2244006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2244006 5 g
AAB2244014 25 g
AAB2244022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2244006 Supplier Thermo Scientific Chemicals Supplier No. B2244006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-40-6 | C7H10N2 | 122.171 g/mol

1-Methyl-1-phenylhydrazine is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-1-phenylhydrazine is used to make other chemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 2 - 8°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 618-40-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00007627
InChI Key MWOODERJGVWYJE-UHFFFAOYSA-N
Synonym 1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
PubChem CID 36299
IUPAC Name 1-methyl-1-phenylhydrazine
SMILES CN(C1=CC=CC=C1)N

Specifications

Density 1.038
Boiling Point 54°C to 55°C (0.3 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.583
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 1-Methyl-1-phenylhydrazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-547-9
RTECSNumber MV7712000
TSCA Yes

RUO – Research Use Only