missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexene 98.0+%, TCI America™

Supplier:  TCI America M0200100ML

 View more versions of this product

Catalog No. M0200100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Methyl-1-cyclohexene
Colorless
C7H12
100 mL
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
CC1=CCCCC1
96.173
96.17
Liquid
591-49-1
110°C
MFCD00001548
3295
CTMHWPIWNRWQEG-UHFFFAOYSA-N
1-methylcyclohexene
11574
≥98.0% (GC)

Chemical Identifiers

591-49-1
96.173
CTMHWPIWNRWQEG-UHFFFAOYSA-N
11574
CC1=CCCCC1
C7H12
MFCD00001548
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
1-methylcyclohexene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions