accessibility menu, dialog, popup

UserName

CAS RN 591-49-1

IUPAC Name: 1-methylcyclohex-1-ene
Synonyms: 1-methylcyclohexene
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: CTMHWPIWNRWQEG-UHFFFAOYSA-N
SMILES: CC1=CCCCC1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)