missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexene 98.0+%, TCI America™

Catalog No. p-7132051
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M020025ML 25 mL
M0200100ML 100 mL
M0200500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M020025ML Supplier TCI America Supplier No. M020025ML
Only null left

Chemical Identifiers

CAS 591-49-1
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00001548
InChI Key CTMHWPIWNRWQEG-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
PubChem CID 11574
IUPAC Name 1-methylcyclohexene
SMILES CC1=CCCCC1

Specifications

Color Colorless
Boiling Point 110°C
Quantity 25 mL
UN Number 3295
Formula Weight 96.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-cyclohexene