missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, ≥99.7% (GC), MilliporeSigma™ Supelco™

$228.82 - $228.82

Specifications

CAS 107-98-2
Ignition Point 532°F
Vapor Pressure 10.9 mmHg (25°C)
Vapor Density 3.12 (vs. air)
Density 0.916 g/mL (at 25°C)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019156
View Documents
Merck Emd Millipore
656925MLF
5 mL
Each for $228.82
Only null left
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

107-98-2
10.9 mmHg (25°C)
0.916 g/mL (at 25°C)
Neat
MFCD00004537
Propylene glycol methyl ether; Propyleneglycol monomethyl ether
90.12
Analytical Standard
532°F
3.12 (vs. air)
82°C - closed cup
CH3 CH(OH)CH2 OCH3
118°C to 119°C
Analytical Standard
≥99.7% (GC)
Videos
SDS
Documents

Documents