missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodo-4-(trifluoromethoxy)benzene 98.0+%, TCI America™
$80.16 - $240.46
Chemical Identifiers
| CAS | 103962-05-6 |
|---|---|
| Molecular Formula | C7H4F3IO |
| Molecular Weight (g/mol) | 288.01 |
| MDL Number | MFCD00042411 |
| InChI Key | RTUDBROGOZBBIC-UHFFFAOYSA-N |
| Synonym | 1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene |
| PubChem CID | 2777294 |
| IUPAC Name | 1-iodo-4-(trifluoromethoxy)benzene |
| SMILES | FC(F)(F)OC1=CC=C(I)C=C1 |
Chemical Identifiers
| 103962-05-6 | |
| 288.01 | |
| RTUDBROGOZBBIC-UHFFFAOYSA-N | |
| 2777294 | |
| FC(F)(F)OC1=CC=C(I)C=C1 |
| C7H4F3IO | |
| MFCD00042411 | |
| 1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene | |
| 1-iodo-4-(trifluoromethoxy)benzene |
Specifications
| 103962-05-6 | |
| C7H4F3IO | |
| 5 g | |
| RTUDBROGOZBBIC-UHFFFAOYSA-N | |
| 1-iodo-4-(trifluoromethoxy)benzene | |
| 2777294 | |
| ≥98.0% (GC) | |
| 1-Iodo-4-(trifluoromethoxy)benzene |
| 97°C | |
| MFCD00042411 | |
| 1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene | |
| FC(F)(F)OC1=CC=C(I)C=C1 | |
| 288.01 | |
| 288.01 | |
| Liquid |
Safety and Handling
TSCA : No