missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-(trifluoromethoxy)benzene 98.0+%, TCI America™

Supplier:  TCI America T215325G

 View more versions of this product

Catalog No. T215325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Iodo-4-(trifluoromethoxy)benzene
97°C
MFCD00042411
1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
FC(F)(F)OC1=CC=C(I)C=C1
288.01
288.01
Liquid
103962-05-6
C7H4F3IO
25 g
RTUDBROGOZBBIC-UHFFFAOYSA-N
1-iodo-4-(trifluoromethoxy)benzene
2777294
≥98.0% (GC)

Chemical Identifiers

103962-05-6
288.01
RTUDBROGOZBBIC-UHFFFAOYSA-N
2777294
FC(F)(F)OC1=CC=C(I)C=C1
C7H4F3IO
MFCD00042411
1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
1-iodo-4-(trifluoromethoxy)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions