missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-n-propylbenzene, 97%

Catalog No. p-7044307
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0576706 5 g
AAL0576714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0576706 Supplier Thermo Scientific Chemicals Supplier No. L0576706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126261-84-5 | C9H11I | 246.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126261-84-5
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.09
MDL Number MFCD00051612
InChI Key ZRWCHMFGMKNWEC-UHFFFAOYSA-N
Synonym 1-iodo-4-n-propylbenzene, 1-iodo-4-propyl-benzene, 4-iodopropylbenzene, 4-propyliodobenzene, p-n-propyliodobenzene, 4-iodo-n-propylbenzene, 4-n-propyl iodobenzene, #, benzene,1-iodo-4-propyl, 1-iodanyl-4-propyl-benzene
PubChem CID 612541
IUPAC Name 1-iodo-4-propylbenzene
SMILES CCCC1=CC=C(I)C=C1

Specifications

Boiling Point 240°C to 242°C
Refractive Index 1.579
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 97%
Chemical Name or Material 1-Iodo-4-n-propylbenzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only