CAS RN 126261-84-5
IUPAC Name:
1-iodo-4-propylbenzene
Synonyms:
1-iodo-4-propylbenzene
Molecular Weight (g/mol):
246.09
Molecular Formula:
C9H11I
InChi Key:
ZRWCHMFGMKNWEC-UHFFFAOYSA-N
SMILES:
CCCC1=CC=C(I)C=C1
1
–
1
of
1
results
1
Promotions Available
1-Iodo-4-n-propylbenzene, 97%
1-Iodo-4-n-propylbenzene, 97%
CAS: 126261-84-5 Molecular Formula: C9H11I Molecular Weight (g/mol): 246.09 MDL Number: MFCD00051612 InChI Key: ZRWCHMFGMKNWEC-UHFFFAOYSA-N Synonym: 1-iodo-4-n-propylbenzene,1-iodo-4-propyl-benzene,4-iodopropylbenzene,4-propyliodobenzene,p-n-propyliodobenzene,4-iodo-n-propylbenzene,4-n-propyl iodobenzene,#,benzene,1-iodo-4-propyl,1-iodanyl-4-propyl-benzene PubChem CID: 612541 IUPAC Name: 1-iodo-4-propylbenzene SMILES: CCCC1=CC=C(I)C=C1