missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-4-nitroanthraquinone 97.0+%, TCI America™

Catalog No. p-7130713
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07655G 5 g
H076525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07655G Supplier TCI America Supplier No. H07655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-65-2
Molecular Formula C14H7NO5
Molecular Weight (g/mol) 269.212
MDL Number MFCD00142642
InChI Key XXUOKOYLVBHBCG-UHFFFAOYSA-N
PubChem CID 3490678
IUPAC Name 1-hydroxy-4-nitroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)[N+](=O)[O-]

Specifications

Melting Point 272°C
Quantity 5 g
Formula Weight 269.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxy-4-nitroanthraquinone
EINECSNumber (4)-0735
TSCA No