accessibility menu, dialog, popup

UserName

CAS RN 81-65-2

IUPAC Name: 1-hydroxy-4-nitro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-hydroxy-4-nitroanthracene-9,10-dione
Molecular Weight (g/mol): 269.21
Molecular Formula: C14H7NO5
InChi Key: XXUOKOYLVBHBCG-UHFFFAOYSA-N
SMILES: OC1=CC=C(C2=C1C(=O)C1=CC=CC=C1C2=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results