missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyne, 98+%

CAS: 693-02-7 | C6H10 | 82.146 g/mol

Supplier:  Thermo Scientific Chemicals A1315618

 View more versions of this product

Catalog No. AAA1315618

Only null left
Explore available promotions

Description

Description

1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Specifications

Specifications

1-Hexyne
-132°C
69°C to 71°C
Characteristic
1.399
50 g
635687
Miscible with organic solvents. Immiscible with water.
CCCCC#C
82.146
82.15
693-02-7
0.718
−21°C (−5°F)
C6H10
MFCD00009504
UN3295
1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
CGHIBGNXEGJPQZ-UHFFFAOYSA-N
hex-1-yne
12732
≥98%

Chemical Identifiers

693-02-7
82.146
CGHIBGNXEGJPQZ-UHFFFAOYSA-N
12732
CCCCC#C
C6H10
MFCD00009504
1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
hex-1-yne
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 211-736-9

RTECSNumber : MQ9691000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only