missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyne, 98+%

Catalog No. p-7021002
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1315609 10 g
AAA1315618 50 g
AAA1315630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1315609 Supplier Thermo Scientific Chemicals Supplier No. A1315609

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 693-02-7 | C6H10 | 82.146 g/mol

1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.

Chemical Identifiers

CAS 693-02-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009504
InChI Key CGHIBGNXEGJPQZ-UHFFFAOYSA-N
Synonym 1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
PubChem CID 12732
IUPAC Name hex-1-yne
SMILES CCCCC#C

Specifications

Melting Point -132°C
Density 0.718
Boiling Point 69°C to 71°C
Flash Point −21°C (−5°F)
Odor Characteristic
Refractive Index 1.399
Quantity 10 g
UN Number UN3295
Beilstein 635687
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 82.15
Percent Purity ≥98%
Chemical Name or Material 1-Hexyne
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-736-9
RTECSNumber MQ9691000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only