accessibility menu, dialog, popup

UserName

CAS RN 693-02-7

IUPAC Name: hex-1-yne
Synonyms: hexyne
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: CGHIBGNXEGJPQZ-UHFFFAOYSA-N
SMILES: CCCCC#C
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)