missing translation for 'onlineSavingsMsg'
Learn More

1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol

CAS: 1629-60-3 | C6H10O | 98.145 g/mol

Supplier:  Thermo Scientific Chemicals L0766014

 View more versions of this product

Catalog No. AAL0766014


Only null left
Explore available promotions

Description

Description

1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water (slightly).

Notes
Store at 4°C. Protect from heat. Incompatible with oxidizing agents, heat, light.
Specifications

Specifications

1-Hexen-3-one
1629-60-3
127°C to 129°C
Pungent
1.4275
25 g
1740796
Soluble in water (slightly).
CCCC(=O)C=C
98.145
98.15
stabilized with 0.5% 4-methoxyphenol
0.84
22°C (71°F)
C6H10O
MFCD00051563
UN1224
1-hexen-3-one, propyl vinyl ketone, unii-qh9d98z86n, 1-hexenone-3, n-propylacrolein, vinyl propyl ketone, acmc-1bt5f, 1-hexene-3-one
JTHNLKXLWOXOQK-UHFFFAOYSA-N
hex-1-en-3-one
15395
≥90%

Chemical Identifiers

1629-60-3
98.145
JTHNLKXLWOXOQK-UHFFFAOYSA-N
15395
CCCC(=O)C=C
C6H10O
MFCD00051563
1-hexen-3-one, propyl vinyl ketone, unii-qh9d98z86n, 1-hexenone-3, n-propylacrolein, vinyl propyl ketone, acmc-1bt5f, 1-hexene-3-one
hex-1-en-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 216-625-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only