missing translation for 'onlineSavingsMsg'
Learn More

1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol

Catalog No. p-7044668
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0766006 5 g
AAL0766014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0766006 Supplier Thermo Scientific Chemicals Supplier No. L0766006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1629-60-3 | C6H10O | 98.145 g/mol

1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water (slightly).

Notes
Store at 4°C. Protect from heat. Incompatible with oxidizing agents, heat, light.

Chemical Identifiers

CAS 1629-60-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00051563
InChI Key JTHNLKXLWOXOQK-UHFFFAOYSA-N
Synonym 1-hexen-3-one, propyl vinyl ketone, unii-qh9d98z86n, 1-hexenone-3, n-propylacrolein, vinyl propyl ketone, acmc-1bt5f, 1-hexene-3-one
PubChem CID 15395
IUPAC Name hex-1-en-3-one
SMILES CCCC(=O)C=C

Specifications

Density 0.84
Boiling Point 127°C to 129°C
Flash Point 22°C (71°F)
Odor Pungent
Refractive Index 1.4275
Quantity 5 g
UN Number UN1224
Beilstein 1740796
Solubility Information Soluble in water (slightly).
Formula Weight 98.15
Percent Purity ≥90%
Chemical Name or Material 1-Hexen-3-one, stabilized with 0.5% 4-methoxyphenol
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 216-625-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only