accessibility menu, dialog, popup

UserName

CAS RN 1629-60-3

IUPAC Name: hex-1-en-3-one
Synonyms: hex-1-en-3-one
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: JTHNLKXLWOXOQK-UHFFFAOYSA-N
SMILES: CCCC(=O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results