missing translation for 'onlineSavingsMsg'
Learn More

1-Heptyloxy-4-nitrobenzene, TCI America™

Catalog No. H04355ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04355ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04355ML Supplier TCI America Supplier No. H04355ML
Only null left

Chemical Identifiers

CAS 13565-36-1
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD00024692
InChI Key AVDKYVSWJDZALX-UHFFFAOYSA-N
Synonym Heptyl 4-Nitrophenyl Ether
PubChem CID 139499
IUPAC Name 1-heptoxy-4-nitrobenzene
SMILES CCCCCCCOC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Boiling Point 203°C
Quantity 5 mL
Formula Weight 237.30
Physical Form Liquid
Chemical Name or Material 1-Heptyloxy-4-nitrobenzene