accessibility menu, dialog, popup

UserName

H3CONOO

CAS RN 13565-36-1

IUPAC Name: 1-(heptyloxy)-4-nitrobenzene
Synonyms: 1-(heptyloxy)-4-nitrobenzene
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: AVDKYVSWJDZALX-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results