missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptanol 98.0+%, TCI America™
$34.18 - $83.54
Chemical Identifiers
| CAS | 111-70-6 |
|---|---|
| Molecular Formula | C7H16O |
| Molecular Weight (g/mol) | 116.20 |
| MDL Number | MFCD00002986 |
| InChI Key | BBMCTIGTTCKYKF-UHFFFAOYSA-N |
| Synonym | 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol |
| PubChem CID | 8129 |
| IUPAC Name | heptan-1-ol |
| SMILES | CCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H003325ML
|
N/A | 25 mL |
Each for $34.18
|
|
|||||
|
H0033500ML
|
N/A | 500 mL |
Each for $83.54
|
|
|||||
Chemical Identifiers
| C7H16O | |
| MFCD00002986 | |
| 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol | |
| heptan-1-ol |
Specifications
| 111-70-6 | |
| Colorless | |
| C7H16O | |
| 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol | |
| CCCCCCCO | |
| 116.20 | |
| 116.20 | |
| Liquid |
| -34°C | |
| 177°C | |
| MFCD00002986 | |
| BBMCTIGTTCKYKF-UHFFFAOYSA-N | |
| heptan-1-ol | |
| 8129 | |
| ≥98.0% (GC) | |
| 1-Heptanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MK0350000
TSCA : Yes