missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol 98.0+%, TCI America™

Catalog No. H003325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H003325ML 25 mL
H0033500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H003325ML Supplier TCI America Supplier No. H003325ML

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Melting Point -34°C
Color Colorless
Boiling Point 177°C
Quantity 25 mL
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Heptanol
EINECSNumber (2)-0217
RTECSNumber MK0350000
TSCA Yes