missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptanol 98,0+%, TCI America™
Specifications
Specifications
| Melting Point | -34°C |
| Color | Incolore |
| Boiling Point | 177°C |
| Quantity | 500 mL |
| Formula Weight | 116.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 1-Heptanol |
Safety and Handling
| EINECSNumber | (2)-0217 |
| RTECSNumber | MK0350000 |
| TSCA | Yes |