missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Fluoro-4-iodobenzene (stabilized with Copper chip) 98.0+%, TCI America™
Supplier: TCI America F023725G
Specifications
| 1-Fluoro-4-iodobenzene (stabilized with Copper chip) | |
| -20°C | |
| 183°C | |
| MFCD00001052 | |
| 4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene | |
| FC1=CC=C(I)C=C1 | |
| 222.00 | |
| 222.00 | |
| Liquid |
| 352-34-1 | |
| Yellow | |
| C6H4FI | |
| 25 g | |
| KGNQDBQYEBMPFZ-UHFFFAOYSA-N | |
| 1-fluoro-4-iodobenzene | |
| 9605 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 352-34-1 | |
| 222.00 | |
| KGNQDBQYEBMPFZ-UHFFFAOYSA-N | |
| 9605 | |
| FC1=CC=C(I)C=C1 |
| C6H4FI | |
| MFCD00001052 | |
| 4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene | |
| 1-fluoro-4-iodobenzene |
Safety and Handling
TSCA : Yes