missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-4-iodobenzene (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7129563
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02375G 5 g
F023725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02375G Supplier TCI America Supplier No. F02375G
Only null left

Chemical Identifiers

CAS 352-34-1
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001052
InChI Key KGNQDBQYEBMPFZ-UHFFFAOYSA-N
Synonym 4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene
PubChem CID 9605
IUPAC Name 1-fluoro-4-iodobenzene
SMILES FC1=CC=C(I)C=C1

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 183°C
Quantity 5 g
Formula Weight 222.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Fluoro-4-iodobenzene (stabilized with Copper chip)