missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-nitrobenzene, 98+%

Catalog No. p-7028312
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2555314 25 g
AAB2555322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2555314 Supplier Thermo Scientific Chemicals Supplier No. B2555314

CAS: 402-67-5 | C6H4FNO2 | 141.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-67-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD00007196
InChI Key WMASLRCNNKMRFP-UHFFFAOYSA-N
Synonym 3-fluoronitrobenzene, m-fluoronitrobenzene, m-nitrofluorobenzene, benzene, 1-fluoro-3-nitro, 3-fluoro-1-nitrobenzene, 1-fluoro-3-nitro-benzene, 3-fluoro-nitrobenzene, 3-nitro-fluorobenzene, pubchem2286, 3-fluoro nitro benzene
PubChem CID 9823
IUPAC Name 1-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(F)=C1

Specifications

Melting Point 2°C
Density 1.325
Boiling Point 204°C to 206°C
Flash Point 76°C (168°F)
Refractive Index 1.525
Quantity 25 g
UN Number UN2810
Beilstein 1862210
Formula Weight 141.1
Percent Purity ≥98%
Chemical Name or Material 1-Fluoro-3-nitrobenzene
Show More Show Less
Hazard Category H227-H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H331-H373-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-953-0
RTECSNumber DA1385000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only