missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-iodo-5-nitrobenzene 98.0+%, TCI America™

Catalog No. F08151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08151G 1 g
F08155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08151G Supplier TCI America Supplier No. F08151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3819-88-3
Molecular Formula C6H3FINO2
Molecular Weight (g/mol) 266.998
MDL Number MFCD00007219
InChI Key MXPYCSFCKXSPAB-UHFFFAOYSA-N
Synonym 3-fluoro-5-iodonitrobenzene, benzene, 1-fluoro-3-iodo-5-nitro, acmc-1cr1w, 5-fluoro-3-nitroiodobenzene, ksc495i2h, 1-iodo-3-fluoro-5-nitrobenzene, 1-fluoro-3-iodo-5-nitro-benzene, 1-fluoro-3-iodo-5-nitrobenzene
PubChem CID 259086
IUPAC Name 1-fluoro-3-iodo-5-nitrobenzene
SMILES C1=C(C=C(C=C1F)I)[N+](=O)[O-]

Specifications

Melting Point 79°C
Color Yellow
Quantity 1 g
Formula Weight 267.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluoro-3-iodo-5-nitrobenzene