missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Fluoro-2-(trifluoromethoxy)benzene 98.0+%, TCI America™
$57.79 - $164.03
Chemical Identifiers
| CAS | 2106-18-5 |
|---|---|
| Molecular Formula | C7H4F4O |
| Molecular Weight (g/mol) | 180.102 |
| MDL Number | MFCD00236322 |
| InChI Key | UKRYEFFTFFRSPY-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethoxy fluorobenzene, 1-fluoro-2-trifluoromethoxy benzene, 2-fluorotrifluoromethoxybenzene, alpha,alpha,alpha,2-tetrafluoroanisole, benzene, fluoro trifluoromethoxy, 2-fluorophenyl trifluoromethyl ether, trifluoro 2-fluorophenoxy methane, otf-byl, pubchem10392 |
| PubChem CID | 2777283 |
| IUPAC Name | 1-fluoro-2-(trifluoromethoxy)benzene |
| SMILES | C1=CC=C(C(=C1)OC(F)(F)F)F |
Chemical Identifiers
| 2106-18-5 | |
| 180.102 | |
| UKRYEFFTFFRSPY-UHFFFAOYSA-N | |
| 2777283 | |
| C1=CC=C(C(=C1)OC(F)(F)F)F |
| C7H4F4O | |
| MFCD00236322 | |
| 2-trifluoromethoxy fluorobenzene, 1-fluoro-2-trifluoromethoxy benzene, 2-fluorotrifluoromethoxybenzene, alpha,alpha,alpha,2-tetrafluoroanisole, benzene, fluoro trifluoromethoxy, 2-fluorophenyl trifluoromethyl ether, trifluoro 2-fluorophenoxy methane, otf-byl, pubchem10392 | |
| 1-fluoro-2-(trifluoromethoxy)benzene |
Specifications
| 2106-18-5 | |
| 105°C | |
| MFCD00236322 | |
| 1993 | |
| UKRYEFFTFFRSPY-UHFFFAOYSA-N | |
| 1-fluoro-2-(trifluoromethoxy)benzene | |
| 2777283 | |
| ≥98.0% (GC) | |
| 1-Fluoro-2-(trifluoromethoxy)benzene |
| Yellow | |
| C7H4F4O | |
| 5 g | |
| 2-trifluoromethoxy fluorobenzene, 1-fluoro-2-trifluoromethoxy benzene, 2-fluorotrifluoromethoxybenzene, alpha,alpha,alpha,2-tetrafluoroanisole, benzene, fluoro trifluoromethoxy, 2-fluorophenyl trifluoromethyl ether, trifluoro 2-fluorophenoxy methane, otf-byl, pubchem10392 | |
| C1=CC=C(C(=C1)OC(F)(F)F)F | |
| 180.102 | |
| 180.10 | |
| Liquid |
Safety and Handling
TSCA : No