accessibility menu, dialog, popup

UserName

FOFFF

CAS RN 2106-18-5

IUPAC Name: 1-fluoro-2-(trifluoromethoxy)benzene
Synonyms: 1-fluoro-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: UKRYEFFTFFRSPY-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1OC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results