missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-nitrobenzene, 99%

Catalog No. p-7021496
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1385218 50 g
AAA1385230 250 g
AAA138520B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1385218 Supplier Thermo Scientific Chemicals Supplier No. A1385218

CAS: 1493-27-2 | C6H4FNO2 | 141.101 g/mol

1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

Solubility
Immiscible in water.

Notes
Avoid excessive heat and light.

Chemical Identifiers

CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD00007048
InChI Key PWKNBLFSJAVFAB-UHFFFAOYSA-N
Synonym 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
PubChem CID 73895
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])F

Specifications

Melting Point °C to 8°C
Density 1.340
Boiling Point 215°C
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.5317
Quantity 50 g
UN Number UN2810
Beilstein 607262
Solubility Information Immiscible in water.
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less
Hazard Category H301-H310-H315-H319-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373-H315-H319
Precautionary Statement P260-P262-P264b-P270-P280-P301+P310-P302+P350-P305+P351+P338-P310-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 216-088-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only