missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-iodobenzene 99.0+%, TCI America™

Catalog No. p-7129572
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F025310G 10 g
F025325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F025310G Supplier TCI America Supplier No. F025310G
Only null left

Chemical Identifiers

CAS 348-52-7
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001031
InChI Key TYHUGKGZNOULKD-UHFFFAOYSA-N
Synonym 2-fluoroiodobenzene, o-fluoroiodobenzene, o-iodofluorobenzene, benzene, 1-fluoro-2-iodo, 2-iodofluorobenzene, 3-fluoro-2-iodobenzene, 1-fluoro-2-iodo-benzene, 2-fluoro-iodobenzene, 2-fluoro-1-iodobenzene
PubChem CID 67673
IUPAC Name 1-fluoro-2-iodobenzene
SMILES FC1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 189°C
Quantity 10 g
Formula Weight 222.00
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-iodobenzene