accessibility menu, dialog, popup

UserName

FI

CAS RN 348-52-7

IUPAC Name: 1-fluoro-2-iodobenzene
Synonyms: benzene, 1-fluoro-2-iodo-
Molecular Weight (g/mol): 222
Molecular Formula: C6H4FI
InChi Key: TYHUGKGZNOULKD-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1I
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)