missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2,4-dimethoxybenzene, 97%

Catalog No. AAH6352803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6352803 1 g
AAH6352806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6352803 Supplier Thermo Scientific Chemicals Supplier No. H6352803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17715-70-7 | C8H9FO2 | 156.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17715-70-7
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00042265
InChI Key QLJNEPOEZGFNEA-UHFFFAOYSA-N
PubChem CID 2758413
IUPAC Name 1-fluoro-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)F)OC

Specifications

Color Yellow
Density 1.188 g/mL
Flash Point 105°C (221°F)
Quantity 1 g
Synonym 2,4-dimethoxy-1-fluorobenzene, 2,4-dimethoxyfluorobenzene, 1,3-dimethoxy-4-fluorobenzene, 4-fluoro-3-methoxyanisole, benzene, 1-fluoro-2,4-dimethoxy, 2,4-dimethoxy-1-fluoro-benzene, acmc-1c4p9, 1-fluoro-2,4-dimethoxy-benzene, qljnepoezgfnea-uhfffaoysa, benzene,1-fluoro-2,4-dimethoxy
Formula Weight 156.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2,4-dimethoxybenzene
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 605-794-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only