accessibility menu, dialog, popup

UserName

H3COFOCH3

CAS RN 17715-70-7

IUPAC Name: 1-fluoro-2,4-dimethoxybenzene
Synonyms: 1-fluoro-2,4-dimethoxybenzene
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: QLJNEPOEZGFNEA-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results