missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynylpyrene 98.0+%, TCI America™
Supplier: TCI America E09391G
Specifications
| 1-Ethynylpyrene | |
| 117°C | |
| MFCD02093933 | |
| 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 | |
| C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 | |
| 226.28 | |
| 226.28 | |
| Crystalline Powder |
| 34993-56-1 | |
| C18H10 | |
| 1 g | |
| VEBUBSLYGRMOSZ-UHFFFAOYSA-N | |
| 1-ethynylpyrene | |
| 154905 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 34993-56-1 | |
| 226.28 | |
| VEBUBSLYGRMOSZ-UHFFFAOYSA-N | |
| 154905 | |
| C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
| C18H10 | |
| MFCD02093933 | |
| 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 | |
| 1-ethynylpyrene |
Safety and Handling
TSCA : No