missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylpyrene 98.0+%, TCI America™

Catalog No. p-7129234
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0939200MG 200 mg
E09391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0939200MG Supplier TCI America Supplier No. E0939200MG
Only null left

Chemical Identifiers

CAS 34993-56-1
Molecular Formula C18H10
Molecular Weight (g/mol) 226.28
MDL Number MFCD02093933
InChI Key VEBUBSLYGRMOSZ-UHFFFAOYSA-N
Synonym 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2
PubChem CID 154905
IUPAC Name 1-ethynylpyrene
SMILES C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 117°C
Quantity 200 mg
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethynylpyrene