missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-propylbenzene 96.0+%, TCI America™

Catalog No. p-7129131
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07505G 5 g
E075025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07505G Supplier TCI America Supplier No. E07505G
Only null left

Chemical Identifiers

CAS 62452-73-7
Molecular Formula C11H12
Molecular Weight (g/mol) 144.217
MDL Number MFCD00173886
InChI Key UVFFOABHOIMLNB-UHFFFAOYSA-N
Synonym 4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
PubChem CID 2775133
IUPAC Name 1-ethynyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C#C

Specifications

Quantity 5 g
Formula Weight 144.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-4-propylbenzene