missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynyl-3-fluorobenzene 98.0+%, TCI America™
$167.76 - $667.33
Chemical Identifiers
| CAS | 2561-17-3 |
|---|---|
| Molecular Formula | C8H5F |
| Molecular Weight (g/mol) | 120.126 |
| MDL Number | MFCD03093798 |
| InChI Key | PTRUTZFCVFUTMW-UHFFFAOYSA-N |
| Synonym | 3-Fluorophenylacetylene |
| PubChem CID | 520218 |
| IUPAC Name | 1-ethynyl-3-fluorobenzene |
| SMILES | C#CC1=CC(=CC=C1)F |
Chemical Identifiers
| 2561-17-3 | |
| 120.126 | |
| PTRUTZFCVFUTMW-UHFFFAOYSA-N | |
| 520218 | |
| C#CC1=CC(=CC=C1)F |
| C8H5F | |
| MFCD03093798 | |
| 3-Fluorophenylacetylene | |
| 1-ethynyl-3-fluorobenzene |
Specifications
| 2561-17-3 | |
| 40°C | |
| MFCD03093798 | |
| 1993 | |
| PTRUTZFCVFUTMW-UHFFFAOYSA-N | |
| 1-ethynyl-3-fluorobenzene | |
| 520218 | |
| ≥98.0% (GC) | |
| 1-Ethynyl-3-fluorobenzene |
| Yellow | |
| C8H5F | |
| 5 g | |
| 3-Fluorophenylacetylene | |
| C#CC1=CC(=CC=C1)F | |
| 120.126 | |
| 120.13 | |
| Liquid |
Safety and Handling
TSCA : No