missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynyl-3-fluorobenzene 98.0+%, TCI America™
Supplier: TCI America E133225G
Specifications
| 1-Ethynyl-3-fluorobenzene | |
| Yellow | |
| C8H5F | |
| 25 g | |
| 3-Fluorophenylacetylene | |
| C#CC1=CC(=CC=C1)F | |
| 120.126 | |
| 120.13 | |
| Liquid |
| 2561-17-3 | |
| 40°C | |
| MFCD03093798 | |
| 1993 | |
| PTRUTZFCVFUTMW-UHFFFAOYSA-N | |
| 1-ethynyl-3-fluorobenzene | |
| 520218 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2561-17-3 | |
| 120.126 | |
| PTRUTZFCVFUTMW-UHFFFAOYSA-N | |
| 520218 | |
| C#CC1=CC(=CC=C1)F |
| C8H5F | |
| MFCD03093798 | |
| 3-Fluorophenylacetylene | |
| 1-ethynyl-3-fluorobenzene |
Safety and Handling
TSCA : No