missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylpyrrolidine 98.0+%, TCI America™
$109.98 - $328.08
Chemical Identifiers
| CAS | 7335-06-0 |
|---|---|
| Molecular Formula | C6H13N |
| Molecular Weight (g/mol) | 99.177 |
| MDL Number | MFCD01632186 |
| InChI Key | ONQBOTKLCMXPOF-UHFFFAOYSA-N |
| PubChem CID | 81782 |
| IUPAC Name | 1-ethylpyrrolidine |
| SMILES | CCN1CCCC1 |
Chemical Identifiers
| 7335-06-0 | |
| 99.177 | |
| ONQBOTKLCMXPOF-UHFFFAOYSA-N | |
| 1-ethylpyrrolidine |
| C6H13N | |
| MFCD01632186 | |
| 81782 | |
| CCN1CCCC1 |
Specifications
| 7335-06-0 | |
| 105°C | |
| MFCD01632186 | |
| 2733 | |
| CCN1CCCC1 | |
| 99.177 | |
| 99.18 | |
| Liquid |
| Yellow | |
| C6H13N | |
| 5 g | |
| ONQBOTKLCMXPOF-UHFFFAOYSA-N | |
| 1-ethylpyrrolidine | |
| 81782 | |
| ≥98.0% (GC) | |
| 1-Ethylpyrrolidine |
Safety and Handling
EINECSNumber : (5)-6747
TSCA : No