missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylpyrrolidine 98.0+%, TCI America™
Supplier: TCI America E05105G
Specifications
| 1-Ethylpyrrolidine | |
| Yellow | |
| C6H13N | |
| 5 g | |
| ONQBOTKLCMXPOF-UHFFFAOYSA-N | |
| 1-ethylpyrrolidine | |
| 81782 | |
| ≥98.0% (GC) |
| 7335-06-0 | |
| 105°C | |
| MFCD01632186 | |
| 2733 | |
| CCN1CCCC1 | |
| 99.177 | |
| 99.18 | |
| Liquid |
Chemical Identifiers
| 7335-06-0 | |
| 99.177 | |
| ONQBOTKLCMXPOF-UHFFFAOYSA-N | |
| 1-ethylpyrrolidine |
| C6H13N | |
| MFCD01632186 | |
| 81782 | |
| CCN1CCCC1 |
Safety and Handling
EINECSNumber : (5)-6747
TSCA : No