accessibility menu, dialog, popup

UserName

CAS RN 7335-06-0

H3CN

CAS RN 7335-06-0

IUPAC Name: 1-ethylpyrrolidine
Synonyms: 1-ethyl-pyrrolidine
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: ONQBOTKLCMXPOF-UHFFFAOYSA-N
SMILES: CCN1CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results