missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3,5-difluorobenzene 98.0+%, TCI America™
Supplier: TCI America E122925G
Specifications
| 1-Ethyl-3,5-difluorobenzene | |
| Yellow | |
| MFCD10000628 | |
| 1993 | |
| CCC1=CC(=CC(=C1)F)F | |
| 142.149 | |
| 142.15 | |
| Liquid |
| 117358-52-8 | |
| C8H8F2 | |
| 25 g | |
| PFINYKRPUJCMCT-UHFFFAOYSA-N | |
| 1-ethyl-3,5-difluorobenzene | |
| 20352892 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 117358-52-8 | |
| 142.149 | |
| PFINYKRPUJCMCT-UHFFFAOYSA-N | |
| 1-ethyl-3,5-difluorobenzene |
| C8H8F2 | |
| MFCD10000628 | |
| 20352892 | |
| CCC1=CC(=CC(=C1)F)F |
Safety and Handling
TSCA : No