accessibility menu, dialog, popup

UserName

H3CFF

CAS RN 117358-52-8

IUPAC Name: 1-ethyl-3,5-difluorobenzene
Synonyms: 1-ethyl-3,5-difluorobenzene
Molecular Weight (g/mol): 142.15
Molecular Formula: C8H8F2
InChi Key: PFINYKRPUJCMCT-UHFFFAOYSA-N
SMILES: CCC1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results