missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3,5-difluorobenzene 98.0+%, TCI America™

Catalog No. E12295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12295G 5 g
E122925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12295G Supplier TCI America Supplier No. E12295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117358-52-8
Molecular Formula C8H8F2
Molecular Weight (g/mol) 142.149
MDL Number MFCD10000628
InChI Key PFINYKRPUJCMCT-UHFFFAOYSA-N
PubChem CID 20352892
IUPAC Name 1-ethyl-3,5-difluorobenzene
SMILES CCC1=CC(=CC(=C1)F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 142.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3,5-difluorobenzene